missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-phenylthiophene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 85°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | thiophene,2-bromo-5-phenyl, 2-bromo-5-phenyl-thiophene, 2-phenyl-5-bromothiophene, thiophene, 2-bromo-5-phenyl, 5-bromo-2-phenylthiophene, acmc-209h9c, 2-bromo-5-phenyl thiophene |
| Formula Weight | 239.13 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Bromo-5-phenylthiophene |
Safety and Handling
| TSCA | No |