missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-nitrotoluene 98.0+%, TCI America™
$152.86 - $555.49
Chemical Identifiers
| CAS | 7149-70-4 |
|---|---|
| Molecular Formula | C7H6BrNO2 |
| Molecular Weight (g/mol) | 216.034 |
| MDL Number | MFCD00007281 |
| InChI Key | HIMGPQVBNICCGL-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene |
| PubChem CID | 251672 |
| IUPAC Name | 1-bromo-2-methyl-4-nitrobenzene |
| SMILES | CC1=C(C=CC(=C1)[N+](=O)[O-])Br |
Chemical Identifiers
| 7149-70-4 | |
| 216.034 | |
| HIMGPQVBNICCGL-UHFFFAOYSA-N | |
| 251672 | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])Br |
| C7H6BrNO2 | |
| MFCD00007281 | |
| 2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene | |
| 1-bromo-2-methyl-4-nitrobenzene |
Specifications
| 7149-70-4 | |
| 143°C | |
| MFCD00007281 | |
| 2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])Br | |
| 216.034 | |
| 216.03 | |
| Crystalline Powder |
| 78°C | |
| C7H6BrNO2 | |
| 5 g | |
| HIMGPQVBNICCGL-UHFFFAOYSA-N | |
| 1-bromo-2-methyl-4-nitrobenzene | |
| 251672 | |
| ≥98.0% (GC) | |
| 2-Bromo-5-nitrotoluene |
Safety and Handling
TSCA : No