missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrothiophene, 97%

Catalog No. AC451150050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC451150050 5 g Glass bottle
AC451150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451150050 Supplier Thermo Scientific Chemicals Supplier No. 451150050

CAS: 13195-50-1 | C4H2BrNO2S | 208.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13195-50-1
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.03
InChI Key ZPNFMDYBAQDFDY-UHFFFAOYSA-N
PubChem CID 83222
IUPAC Name 2-bromo-5-nitrothiophene
SMILES C1=C(SC(=C1)Br)[N+](=O)[O-]

Specifications

Melting Point 44°C to 48°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-bromo-2-nitrothiophene, thiophene, 2-bromo-5-nitro, pubchem5283, acmc-1bsij, 2-nitro-5-bromothiophene, 5-nitro-2-bromothiophene, 5-bromo-2-nitro-thiophene, 2-bromo-5-nitro-thiophene, labotest-bb lt00455100, thiophene,2-bromo-5-nitro
Formula Weight 208.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-nitrothiophene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 236-155-8

RUO – Research Use Only