missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroimidazole, 98%

Catalog No. AAH6115503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6115503 1 g
AAH6115506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6115503 Supplier Thermo Scientific Chemicals Supplier No. H6115503

CAS: 65902-59-2 | C3H2BrN3O2 | 191.972 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65902-59-2
Molecular Formula C3H2BrN3O2
Molecular Weight (g/mol) 191.972
MDL Number MFCD09038915
InChI Key UWRJWMLKEHRGOH-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr
PubChem CID 47754
IUPAC Name 2-bromo-5-nitro-1H-imidazole
SMILES C1=C(NC(=N1)Br)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 191.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-nitroimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only