accessibility menu, dialog, popup

UserName

CAS RN 65902-59-2

IUPAC Name: 2-bromo-5-nitro-1H-imidazole
Synonyms: 2-bromo-4-nitro-3H-imidazole
Molecular Weight (g/mol): 191.97
Molecular Formula: C3H2BrN3O2
InChi Key: UWRJWMLKEHRGOH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Br)N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)