missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitroanisole, 98%

Catalog No. p-7028173
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2533104 2 g
AAB2533109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2533104 Supplier Thermo Scientific Chemicals Supplier No. B2533104

CAS: 77337-82-7 | C7H6BrNO3 | 232.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77337-82-7
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00041250
InChI Key NTKADLOYTKVXQN-UHFFFAOYSA-N
Synonym 2-bromo-5-nitroanisole, 4-bromo-3-methoxynitrobenzene, 2-bromo-5-nitroanisol, benzene, 1-bromo-2-methoxy-4-nitro, 1-bromo-2-methoxy-4-nitro-benzene, zlchem 673, pubchem2536, 2-bromo 5-nitro anisole, 2-bromo-5-nitro-anisole, 2-bromo-5-nitro anisole
PubChem CID 101293
IUPAC Name 1-bromo-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 102°C to 107°C
Quantity 2 g
Beilstein 2520496
Formula Weight 232.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitroanisole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 278-669-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only