missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-methylbenzotrifluoride, 98%, Thermo Scientific Chemicals
$146.68 - $146.68
Chemical Identifiers
| CAS | 261952-20-9 |
|---|---|
| Molecular Formula | C8H6BrF3 |
| Molecular Weight (g/mol) | 239.035 |
| MDL Number | MFCD01631586 |
| InChI Key | WXXDBPIOFOYRLP-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl |
| PubChem CID | 2779339 |
| IUPAC Name | 1-bromo-4-methyl-2-(trifluoromethyl)benzene |
| SMILES | CC1=CC(=C(C=C1)Br)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3152303
|
Thermo Scientific Chemicals
H3152303 |
1 g |
Each for $146.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 261952-20-9 | |
| 239.035 | |
| WXXDBPIOFOYRLP-UHFFFAOYSA-N | |
| 2779339 | |
| CC1=CC(=C(C=C1)Br)C(F)(F)F |
| C8H6BrF3 | |
| MFCD01631586 | |
| 2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl | |
| 1-bromo-4-methyl-2-(trifluoromethyl)benzene |
Specifications
| 261952-20-9 | |
| MFCD01631586 | |
| 2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl | |
| CC1=CC(=C(C=C1)Br)C(F)(F)F | |
| 239.035 | |
| 239.04 | |
| 2-Bromo-5-methylbenzotrifluoride |
| C8H6BrF3 | |
| 1 g | |
| WXXDBPIOFOYRLP-UHFFFAOYSA-N | |
| 1-bromo-4-methyl-2-(trifluoromethyl)benzene | |
| 2779339 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only