missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123378
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41455G 5 g
B414525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41455G Supplier TCI America Supplier No. B41455G

Chemical Identifiers

CAS 6967-82-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00079722
InChI Key ZXMISUUIYPFORW-UHFFFAOYSA-N
Synonym 2-bromo-5-methyl-benzoic acid, 6-bromo-m-toluic acid, 2-bromo-5-methyl benzoic acid, benzoic acid, 2-bromo-5-methyl, 4-bromo-3-carboxytoluene, pubchem2166, acmc-1b7tl, ksc357q9r, 2-bromanyl-5-methyl-benzoic acid, 2-bromo-5-methylbenzoic acid
PubChem CID 228080
IUPAC Name 2-bromo-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methylbenzoic Acid