missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylbenzoic acid, 98%

Catalog No. AAH3292703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3292703 1 g
AAH3292706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3292703 Supplier Thermo Scientific Chemicals Supplier No. H3292703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6967-82-4 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6967-82-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00079722
InChI Key ZXMISUUIYPFORW-UHFFFAOYSA-N
Synonym 2-bromo-5-methyl-benzoic acid, 6-bromo-m-toluic acid, 2-bromo-5-methyl benzoic acid, benzoic acid, 2-bromo-5-methyl, 4-bromo-3-carboxytoluene, pubchem2166, acmc-1b7tl, ksc357q9r, 2-bromanyl-5-methyl-benzoic acid, 2-bromo-5-methylbenzoic acid
PubChem CID 228080
IUPAC Name 2-bromo-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Quantity 1 g
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only