missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxytoluene 97.0+%, TCI America™

Catalog No. B178225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B178225G 25 g
B1782250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B178225G Supplier TCI America Supplier No. B178225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27060-75-9
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00060687
InChI Key BLZNSXFQRKVFRP-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
PubChem CID 117915
IUPAC Name 1-bromo-4-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)Br

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 201.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-methoxytoluene
EINECSNumber (3)-4228
TSCA Yes