missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7123527
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44235G 5 g
B442325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44235G Supplier TCI America Supplier No. B44235G

Chemical Identifiers

CAS 19614-12-1
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.959
MDL Number MFCD00078664
InChI Key MURVUTUZSUEIGI-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole
PubChem CID 3525951
IUPAC Name 1-bromo-2-(bromomethyl)-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)CBr

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 279.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methoxybenzyl Bromide