missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzyl bromide, 97%

Catalog No. p-7056188
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6496306 5 g
AAH6496314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6496306 Supplier Thermo Scientific Chemicals Supplier No. H6496306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19614-12-1 | C8H8Br2O | 279.959 g/mol

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 19614-12-1
Molecular Formula C8H8Br2O
Molecular Weight (g/mol) 279.959
MDL Number MFCD00078664
InChI Key MURVUTUZSUEIGI-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzyl bromide, 1-bromo-2-bromomethyl-4-methoxybenzene, 2-bromo-5-methoxybenzylbromide, 4-bromo-3-bromomethyl anisole, 2-bromo-5-methoxy-benzyl bromide, benzene, 1-bromo-2-bromomethyl-4-methoxy, 4-bromo-3-bromomethyl-1-methoxybenzene, pubchem18176, acmc-20a5x2, 4-bromo3-bromomethylanisole
PubChem CID 3525951
IUPAC Name 1-bromo-2-(bromomethyl)-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)CBr

Specifications

Melting Point 90°C to 94°C (literature)
Color White
Quantity 5 g
UN Number UN3261
Beilstein 1945118
Solubility Information Soluble in hot methanol. Slightly soluble in water.
Formula Weight 279.96
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-methoxybenzyl bromide
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only