missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. B38465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38465G 5 g
B384625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38465G Supplier TCI America Supplier No. B38465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22921-68-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00020214
InChI Key ODHJOROUCITYNF-UHFFFAOYSA-N
Synonym 6-bromo-m-anisic acid, 2-bromo-5-methoxybenzoicacid, benzoic acid, 2-bromo-5-methoxy, 5-methoxy-2-bromobenzoic acid, 2-bromo-5-methoxy-benzoic acid, pubchem2674, acmc-1clk6, ksc205k0l, rarechem al be 1111, 2-bromo-5-methoxy benzoic acid
PubChem CID 89906
IUPAC Name 2-bromo-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methoxybenzoic Acid