missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzoic acid, 98+%

Catalog No. AAA1226130
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1226130 250 g
AAA1226109 10 g
AAA1226118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1226130 Supplier Thermo Scientific Chemicals Supplier No. A1226130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22921-68-2 | C8H7BrO3 | 231.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22921-68-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00020214
InChI Key ODHJOROUCITYNF-UHFFFAOYSA-N
Synonym 6-bromo-m-anisic acid, 2-bromo-5-methoxybenzoicacid, benzoic acid, 2-bromo-5-methoxy, 5-methoxy-2-bromobenzoic acid, 2-bromo-5-methoxy-benzoic acid, pubchem2674, acmc-1clk6, ksc205k0l, rarechem al be 1111, 2-bromo-5-methoxy benzoic acid
PubChem CID 89906
IUPAC Name 2-bromo-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Melting Point 156°C to 160°C
Odor Odorless
Quantity 250 g
Beilstein 639763
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-329-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only