missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodobenzoic acid, 97%, Thermo Scientific Chemicals

$390.14 - $390.14

Chemical Identifiers

CAS 25252-00-0
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00079716
InChI Key QPKKBDSNZFSSOD-UHFFFAOYSA-N
Synonym 2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
PubChem CID 2735572
IUPAC Name 2-bromo-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1924706
View Documents
Thermo Scientific Chemicals
L1924706
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25252-00-0
326.915
QPKKBDSNZFSSOD-UHFFFAOYSA-N
2735572
C1=CC(=C(C=C1I)C(=O)O)Br
C7H4BrIO2
MFCD00079716
2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
2-bromo-5-iodobenzoic acid

Specifications

25252-00-0
C7H4BrIO2
5 g
2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)Br
326.915
326.92
2-Bromo-5-iodobenzoic acid
174°C to 176°C
MFCD00079716
Light sensitive
QPKKBDSNZFSSOD-UHFFFAOYSA-N
2-bromo-5-iodobenzoic acid
2735572
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only