missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAL1924706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1924706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1924706 Supplier Thermo Scientific Chemicals Supplier No. L1924706

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25252-00-0
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00079716
InChI Key QPKKBDSNZFSSOD-UHFFFAOYSA-N
Synonym 2-bromo-5-iodobenzoicacid, 2-bromo-5-iodo-benzoic acid, benzoic acid, 2-bromo-5-iodo, pubchem3787, acmc-209gii, intermediates-zcf02680, ksc497i9t, zerenex e/9071914, 2-bromo-5-iodobenzoic acid
PubChem CID 2735572
IUPAC Name 2-bromo-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)Br

Specifications

Melting Point 174°C to 176°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 326.92
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only