missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorotoluene 98.0+%, TCI America™

Catalog No. p-7118983
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13035G 5 g
B130325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13035G Supplier TCI America Supplier No. B13035G
Only null left

Chemical Identifiers

CAS 452-63-1
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00017921
InChI Key RJPNVPITBYXBNB-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene, 1-bromo-4-fluoro-2-methyl-benzene, pubchem1603, 2-bromo-5fluorotoluene, acmc-1acnn, 2-bromo-5-fluoro-toluene
PubChem CID 96743
IUPAC Name 1-bromo-4-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)F)Br

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 g
UN Number 1993
Formula Weight 189.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-fluorotoluene