missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorotoluene, 98+%

Catalog No. p-7081226
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1111214 25 g
AAA1111222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1111214 Supplier Thermo Scientific Chemicals Supplier No. A1111214
Only null left

CAS: 452-63-1 | C7H6BrF | 189.027 g/mol

2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorotoluene is used to produce other chemicals. It can produce 2-Bromo-5-fluoro-benzoic acid. The reaction occurs with reagent KMnO4, solvent H2O and other condition of heating.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 452-63-1
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00017921
InChI Key RJPNVPITBYXBNB-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorotoluene, 3-fluoro-6-bromotoluene, 2-bromo-5-fluoro toluene, benzene, 1-bromo-4-fluoro-2-methyl, 5-fluoro-2-bromotoluene, 1-bromo-4-fluoro-2-methyl-benzene, pubchem1603, 2-bromo-5fluorotoluene, acmc-1acnn, 2-bromo-5-fluoro-toluene
PubChem CID 96743
IUPAC Name 1-bromo-4-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)F)Br

Specifications

Density 1.495
Boiling Point 169°C to 170°C
Flash Point 70°C (158°F)
Refractive Index 1.526
Quantity 25 g
Beilstein 1859121
Solubility Information Difficult to mix.
Formula Weight 189.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorotoluene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 207-203-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only