missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluorophenol 98.0+%, TCI America™
$127.07 - $423.54
Chemical Identifiers
| CAS | 147460-41-1 |
|---|---|
| Molecular Formula | C6H4BrFO |
| Molecular Weight (g/mol) | 190.999 |
| MDL Number | MFCD00040939 |
| InChI Key | HUVAOAVBKOVPBZ-UHFFFAOYSA-N |
| Synonym | phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 |
| PubChem CID | 2724600 |
| IUPAC Name | 2-bromo-5-fluorophenol |
| SMILES | C1=CC(=C(C=C1F)O)Br |
Chemical Identifiers
| 147460-41-1 | |
| 190.999 | |
| HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
| 2724600 | |
| C1=CC(=C(C=C1F)O)Br |
| C6H4BrFO | |
| MFCD00040939 | |
| phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
| 2-bromo-5-fluorophenol |
Specifications
| 147460-41-1 | |
| MFCD00040939 | |
| phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843 | |
| C1=CC(=C(C=C1F)O)Br | |
| 190.999 | |
| 191.00 | |
| Liquid |
| C6H4BrFO | |
| 5 g | |
| HUVAOAVBKOVPBZ-UHFFFAOYSA-N | |
| 2-bromo-5-fluorophenol | |
| 2724600 | |
| ≥98.0% (GC) | |
| 2-Bromo-5-fluorophenol |
Safety and Handling
TSCA : No