missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorophenol 98.0+%, TCI America™

Catalog No. B20405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20405G 5 g
B204025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20405G Supplier TCI America Supplier No. B20405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147460-41-1
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00040939
InChI Key HUVAOAVBKOVPBZ-UHFFFAOYSA-N
Synonym phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843
PubChem CID 2724600
IUPAC Name 2-bromo-5-fluorophenol
SMILES C1=CC(=C(C=C1F)O)Br

Specifications

Quantity 5 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-fluorophenol