missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorophenol, 97%

Catalog No. AAA1506406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1506406 5 g
AAA1506414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1506406 Supplier Thermo Scientific Chemicals Supplier No. A1506406

CAS: 147460-41-1 | C6H4BrFO | 190.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147460-41-1
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00040939
InChI Key HUVAOAVBKOVPBZ-UHFFFAOYSA-N
Synonym phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843
PubChem CID 2724600
IUPAC Name 2-bromo-5-fluorophenol
SMILES C1=CC(=C(C=C1F)O)Br

Specifications

Density 1.717
Boiling Point 184°C to 186°C
Flash Point 87°C (188°F)
Refractive Index 1.554
Quantity 5 g
Beilstein 7915214
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorophenol
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only