missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzonitrile 97.0+%, TCI America™

Catalog No. p-7119891
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30105G 5 g
B301025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30105G Supplier TCI America Supplier No. B30105G
Only null left

Chemical Identifiers

CAS 57381-39-2
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00142875
InChI Key MDHNVHCZDCSTMS-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzenecarbonitrile, 2-bromo-5-fluoro-benzonitrile, benzonitrile, 2-bromo-5-fluoro, 2-cyano-4-fluorobromobenzene, pubchem3218, bromofluorobenzonitrile2, acmc-1awgr, brc6h3 f cn, ksc496i7h, 2-bromo-5-fluorobenzonitrile
PubChem CID 93654
IUPAC Name 2-bromo-5-fluorobenzonitrile
SMILES FC1=CC=C(Br)C(=C1)C#N

Specifications

Melting Point 90°C
Quantity 5 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-fluorobenzonitrile