missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzonitrile, 98+%

Catalog No. p-7027922
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2498906 5 g
AAB2498914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2498906 Supplier Thermo Scientific Chemicals Supplier No. B2498906

CAS: 57381-39-2 | C7H3BrFN | 200.01 g/mol

2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 57381-39-2
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00142875
InChI Key MDHNVHCZDCSTMS-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzenecarbonitrile, 2-bromo-5-fluoro-benzonitrile, benzonitrile, 2-bromo-5-fluoro, 2-cyano-4-fluorobromobenzene, pubchem3218, bromofluorobenzonitrile2, acmc-1awgr, brc6h3 f cn, ksc496i7h, 2-bromo-5-fluorobenzonitrile
PubChem CID 93654
IUPAC Name 2-bromo-5-fluorobenzonitrile
SMILES FC1=CC=C(Br)C(=C1)C#N

Specifications

Melting Point 88°C to 92°C
Quantity 5 g
UN Number UN3439
Beilstein 8678330
Solubility Information Soluble in methanol.
Formula Weight 200.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 260-711-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only