missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluoroanisole 95.0+%, TCI America™
$206.90 - $823.90
Chemical Identifiers
| CAS | 450-88-4 |
|---|---|
| Molecular Formula | C7H6BrFO |
| Molecular Weight (g/mol) | 205.03 |
| MDL Number | MFCD04973752 |
| InChI Key | KGYXKRGMSUHYCY-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-fluoroanisole, benzene, 1-bromo-4-fluoro-2-methoxy, 1-bromo-4-fluoro-2-methoxy-benzene, 2-methoxy-4-fluorobromobenzene, buttpark 87\07-62, pubchem4091, acmc-1ct2m, ksc494a1p, bromo-4-fluoro-2-methoxybenzene |
| PubChem CID | 7018043 |
| IUPAC Name | 1-bromo-4-fluoro-2-methoxybenzene |
| SMILES | COC1=CC(F)=CC=C1Br |
Chemical Identifiers
| 450-88-4 | |
| 205.03 | |
| KGYXKRGMSUHYCY-UHFFFAOYSA-N | |
| 7018043 | |
| COC1=CC(F)=CC=C1Br |
| C7H6BrFO | |
| MFCD04973752 | |
| 2-bromo-5-fluoroanisole, benzene, 1-bromo-4-fluoro-2-methoxy, 1-bromo-4-fluoro-2-methoxy-benzene, 2-methoxy-4-fluorobromobenzene, buttpark 87\07-62, pubchem4091, acmc-1ct2m, ksc494a1p, bromo-4-fluoro-2-methoxybenzene | |
| 1-bromo-4-fluoro-2-methoxybenzene |
Specifications
| 450-88-4 | |
| 208°C | |
| MFCD04973752 | |
| 2-bromo-5-fluoroanisole, benzene, 1-bromo-4-fluoro-2-methoxy, 1-bromo-4-fluoro-2-methoxy-benzene, 2-methoxy-4-fluorobromobenzene, buttpark 87\07-62, pubchem4091, acmc-1ct2m, ksc494a1p, bromo-4-fluoro-2-methoxybenzene | |
| COC1=CC(F)=CC=C1Br | |
| 205.03 | |
| 205.03 | |
| Liquid |
| Yellow | |
| C7H6BrFO | |
| 5 g | |
| KGYXKRGMSUHYCY-UHFFFAOYSA-N | |
| 1-bromo-4-fluoro-2-methoxybenzene | |
| 7018043 | |
| ≥95.0% (GC) | |
| 2-Bromo-5-fluoroanisole |
Safety and Handling
TSCA : No