missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-fluoroanisole 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 208°C |
| Quantity | 25 g |
| Formula Weight | 205.03 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Bromo-5-fluoroanisole |
Safety and Handling
| TSCA | No |