missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoroanisole, 97%

Catalog No. AAH2632506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2632506 5 g
AAH2632514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2632506 Supplier Thermo Scientific Chemicals Supplier No. H2632506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 450-88-4 | C7H6BrFO | 205.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 450-88-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD04973752
InChI Key KGYXKRGMSUHYCY-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroanisole, benzene, 1-bromo-4-fluoro-2-methoxy, 1-bromo-4-fluoro-2-methoxy-benzene, 2-methoxy-4-fluorobromobenzene, buttpark 87\07-62, pubchem4091, acmc-1ct2m, ksc494a1p, bromo-4-fluoro-2-methoxybenzene
PubChem CID 7018043
IUPAC Name 1-bromo-4-fluoro-2-methoxybenzene
SMILES COC1=CC(F)=CC=C1Br

Specifications

Density 1.599
Boiling Point 143°C to 145°C
Flash Point 90°C (194°F)
Refractive Index 1.546
Quantity 5 g
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluoroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only