missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoroaniline 98.0+%, TCI America™

Catalog No. p-7119518
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21635G 5 g
B216325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21635G Supplier TCI America Supplier No. B21635G
Only null left

Chemical Identifiers

CAS 1003-99-2
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00070750
InChI Key FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzenamine, 2-bromo-5-fluoro-aniline, 2-bromo-5-fluoro-phenylamine, 2-bromo-5-fluoro-benzenamine, 2-bromo-5-fluorophenylamine, pubchem1524, 5-fluoro-2-bromoaniline, 2-bromo-5-fluoro aniline, acmc-1c3a5
PubChem CID 2773317
IUPAC Name 2-bromo-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Br

Specifications

Melting Point 45°C
Boiling Point 154°C
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-fluoroaniline