missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-chlorothiophene 95.0+%, TCI America™
$134.21 - $527.52
Chemical Identifiers
| CAS | 2873-18-9 |
|---|---|
| Molecular Formula | C4H2BrClS |
| Molecular Weight (g/mol) | 197.47 |
| MDL Number | MFCD00014523 |
| InChI Key | ZFAJPWYXLYGUJU-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t |
| PubChem CID | 76133 |
| IUPAC Name | 2-bromo-5-chlorothiophene |
| SMILES | ClC1=CC=C(Br)S1 |
Chemical Identifiers
| 2873-18-9 | |
| 197.47 | |
| ZFAJPWYXLYGUJU-UHFFFAOYSA-N | |
| 76133 | |
| ClC1=CC=C(Br)S1 |
| C4H2BrClS | |
| MFCD00014523 | |
| 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t | |
| 2-bromo-5-chlorothiophene |
Specifications
| 2873-18-9 | |
| C4H2BrClS | |
| 5 g | |
| ZFAJPWYXLYGUJU-UHFFFAOYSA-N | |
| 2-bromo-5-chlorothiophene | |
| 76133 | |
| ≥95.0% (GC) | |
| 2-Bromo-5-chlorothiophene |
| 70°C | |
| MFCD00014523 | |
| 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t | |
| ClC1=CC=C(Br)S1 | |
| 197.47 | |
| 197.47 | |
| Liquid |
Safety and Handling
TSCA : No