missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorothiophene, 97%

Catalog No. AAA1668206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1668206 5 g
AAA1668214 25 g
AAA1668222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1668206 Supplier Thermo Scientific Chemicals Supplier No. A1668206

CAS: 2873-18-9 | C4H2BrClS | 197.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2873-18-9
Molecular Formula C4H2BrClS
Molecular Weight (g/mol) 197.47
MDL Number MFCD00014523
InChI Key ZFAJPWYXLYGUJU-UHFFFAOYSA-N
Synonym 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t
PubChem CID 76133
IUPAC Name 2-bromo-5-chlorothiophene
SMILES ClC1=CC=C(Br)S1

Specifications

Density 1.803
Boiling Point 69°C to 70°C (18 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.596
Quantity 5 g
UN Number UN2810
Beilstein 109897
Formula Weight 197.48
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-chlorothiophene
Hazard Category H227-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 220-708-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only