missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7122901
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B33665G 5 g
B336625G 25 g
2 options

Catalog No. B33665G

Supplier: TCI America B33665G

Only null left

Chemical Identifiers

CAS 344-65-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00010308
InChI Key OSTIALFVJOFNPP-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzotrifluoride, 1-bromo-4-chloro-2-trifluoromethyl benzene, benzene, 1-bromo-4-chloro-2-trifluoromethyl, 2-bromo-5-chorobenzotrifluoride, 2-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, 1-bromo-4-chloro-2-trifluoromethyl-benzene, 4-chloro-2-trifluoromethyl bromobenzene, pubchem2713, bromo-5-chlorobenzotrifluoride
PubChem CID 136172
IUPAC Name 1-bromo-4-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Br

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 198°C
Quantity 5 g
Formula Weight 259.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-chlorobenzotrifluoride