missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. B44921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44921G 1 g
B44925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44921G Supplier TCI America Supplier No. B44921G

Chemical Identifiers

CAS 81107-97-3
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD06657954
InChI Key DTEDKIRYMYDIGO-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl phenol, 3-bromo-4-hydroxybenzotrifluoride, phenol, 2-bromo-4-trifluoromethyl, buttpark 180\07-84, pubchem18455, acmc-209pkp, ksc495c0j, 4-trifluoromethyl-2-bromophenol, 2-bromo-4-trifluoromethyl-phenol, phenol,2-bromo-4-trifluoromethyl
PubChem CID 2761168
IUPAC Name 2-bromo-4-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 241.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)phenol