missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)phenol, 98%

CAS: 81107-97-3 | C7H4BrF3O | 241.007 g/mol

$90.19 - $282.45

Chemical Identifiers

CAS 81107-97-3
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD06657954
InChI Key DTEDKIRYMYDIGO-UHFFFAOYSA-N
PubChem CID 2761168
IUPAC Name 2-bromo-4-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6183603
View Documents
Thermo Scientific Chemicals
H6183603
1 g
Each for $90.19
Only null left
 
AAH6183606
View Documents
Thermo Scientific Chemicals
H6183606
5 g
Each for $282.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81107-97-3
241.007
DTEDKIRYMYDIGO-UHFFFAOYSA-N
2-bromo-4-(trifluoromethyl)phenol
C7H4BrF3O
MFCD06657954
2761168
C1=CC(=C(C=C1C(F)(F)F)Br)O

Specifications

81107-97-3
C7H4BrF3O
1 g
2-bromo-4-trifluoromethyl phenol, 3-bromo-4-hydroxybenzotrifluoride, phenol, 2-bromo-4-trifluoromethyl, buttpark 180\07-84, pubchem18455, acmc-209pkp, ksc495c0j, 4-trifluoromethyl-2-bromophenol, 2-bromo-4-trifluoromethyl-phenol, phenol,2-bromo-4-trifluoromethyl
C1=CC(=C(C=C1C(F)(F)F)Br)O
241.007
241.01
Liquid
Brown
MFCD06657954
7702274
DTEDKIRYMYDIGO-UHFFFAOYSA-N
2-bromo-4-(trifluoromethyl)phenol
2761168
98%
2-Bromo-4-(trifluoromethyl)phenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only