missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(tert-butyl)aniline 98.0+%, TCI America™

Catalog No. p-7124041
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B53255G 5 g
B532525G 25 g
2 options

Catalog No. B53255G

Supplier: TCI America B53255G

Only null left

Chemical Identifiers

CAS 103273-01-4
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD00052693
InChI Key OLKYFBNIFKQRIZ-UHFFFAOYSA-N
PubChem CID 238591
IUPAC Name 2-bromo-4-tert-butylaniline
SMILES CC(C)(C)C1=CC(Br)=C(N)C=C1

Specifications

Boiling Point 265°C
Quantity 5 g
Formula Weight 228.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-(tert-butyl)aniline