missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-pyridinecarboxaldehyde 95.0+%, TCI America™

Catalog No. B37151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37151G 1 g
B37155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37151G Supplier TCI America Supplier No. B37151G

Chemical Identifiers

CAS 118289-17-1
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD04039311
InChI Key RTWLIQFKXMWEJY-UHFFFAOYSA-N
Synonym 2-bromopyridine-4-carboxaldehyde, 2-bromo-4-pyridinecarboxaldehyde, 2-bromoisonicotinaldehyde, 2-bromo-4-formylpyridine, 2-bromo-4-pyridinecarbaldehyde, 4-pyridinecarboxaldehyde, 2-bromo, 2-bromopyridine-4-formylaldehyde, pubchem5075, 2-bromisonicotinaldehyd, acmc-2099wk
PubChem CID 2762991
IUPAC Name 2-bromopyridine-4-carbaldehyde
SMILES C1=CN=C(C=C1C=O)Br

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-4-pyridinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator