accessibility menu, dialog, popup

UserName

BrON

CAS RN 118289-17-1

IUPAC Name: 2-bromopyridine-4-carbaldehyde
Synonyms: 2-bromopyridine-4-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: RTWLIQFKXMWEJY-UHFFFAOYSA-N
SMILES: BrC1=CC(C=O)=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results