missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-nitrotoluene 98.0+%, TCI America™
Supplier: TCI America B1262250G
Specifications
| 2-Bromo-4-nitrotoluene | |
| 77°C | |
| C7H6BrNO2 | |
| 250 g | |
| XFZFJQHXWJIBQV-UHFFFAOYSA-N | |
| 2-bromo-1-methyl-4-nitrobenzene | |
| 82189 | |
| ≥98.0% (GC) |
| 7745-93-9 | |
| 175°C | |
| MFCD00007195 | |
| 2-bromo-4-nitrotoluene, benzene, 2-bromo-1-methyl-4-nitro, 3-bromo-4-methylnitrobenzene, toluene, 2-bromo-4-nitro, 3-bromo-4-methyl nitrobenzene, 1-bromo-2-methyl-5-nitrobenzene, 2-bromo-1-methyl-4-nitro-benzene, pubchem3822, 2-bromo-4nitrotoluene, 2-bromo4-nitrotoluene | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])Br | |
| 216.034 | |
| 216.03 | |
| Crystalline Powder |
Chemical Identifiers
| 7745-93-9 | |
| 216.034 | |
| XFZFJQHXWJIBQV-UHFFFAOYSA-N | |
| 82189 | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])Br |
| C7H6BrNO2 | |
| MFCD00007195 | |
| 2-bromo-4-nitrotoluene, benzene, 2-bromo-1-methyl-4-nitro, 3-bromo-4-methylnitrobenzene, toluene, 2-bromo-4-nitro, 3-bromo-4-methyl nitrobenzene, 1-bromo-2-methyl-5-nitrobenzene, 2-bromo-1-methyl-4-nitro-benzene, pubchem3822, 2-bromo-4nitrotoluene, 2-bromo4-nitrotoluene | |
| 2-bromo-1-methyl-4-nitrobenzene |
Safety and Handling
TSCA : No