missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-nitrophenol 98.0+%, TCI America™
$136.08 - $495.84
Chemical Identifiers
| CAS | 5847-59-6 |
|---|---|
| Molecular Formula | C6H4BrNO3 |
| Molecular Weight (g/mol) | 218.006 |
| MDL Number | MFCD06656567 |
| InChI Key | DCIPFSYBGTWYCR-UHFFFAOYSA-N |
| Synonym | 3-Bromo-4-hydroxynitrobenzene |
| PubChem CID | 22109 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])Br)O |
Chemical Identifiers
| 5847-59-6 | |
| 218.006 | |
| DCIPFSYBGTWYCR-UHFFFAOYSA-N | |
| 22109 |
| C6H4BrNO3 | |
| MFCD06656567 | |
| 3-Bromo-4-hydroxynitrobenzene | |
| C1=CC(=C(C=C1[N+](=O)[O-])Br)O |
Specifications
| 5847-59-6 | |
| Yellow | |
| MFCD06656567 | |
| 3077 | |
| DCIPFSYBGTWYCR-UHFFFAOYSA-N | |
| 218.006 | |
| 218.01 | |
| Crystalline Powder |
| 112°C | |
| C6H4BrNO3 | |
| 1 g | |
| 3-Bromo-4-hydroxynitrobenzene | |
| C1=CC(=C(C=C1[N+](=O)[O-])Br)O | |
| 22109 | |
| ≥98.0% (GC,T) | |
| 2-Bromo-4-nitrophenol |
Safety and Handling
TSCA : No