missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-nitroimidazole 98.0+%, TCI America™
$98.18 - $315.72
Chemical Identifiers
| CAS | 65902-59-2 |
|---|---|
| Molecular Formula | C3H2BrN3O2 |
| Molecular Weight (g/mol) | 191.972 |
| MDL Number | MFCD09038915 |
| InChI Key | UWRJWMLKEHRGOH-UHFFFAOYSA-N |
| Synonym | 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr |
| PubChem CID | 47754 |
| IUPAC Name | 2-bromo-5-nitro-1H-imidazole |
| SMILES | C1=C(NC(=N1)Br)[N+](=O)[O-] |
Chemical Identifiers
| 65902-59-2 | |
| 191.972 | |
| UWRJWMLKEHRGOH-UHFFFAOYSA-N | |
| 47754 | |
| C1=C(NC(=N1)Br)[N+](=O)[O-] |
| C3H2BrN3O2 | |
| MFCD09038915 | |
| 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr | |
| 2-bromo-5-nitro-1H-imidazole |
Specifications
| 65902-59-2 | |
| Yellow | |
| MFCD09038915 | |
| 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr | |
| C1=C(NC(=N1)Br)[N+](=O)[O-] | |
| 191.972 | |
| 191.97 | |
| Crystalline Powder |
| 235°C | |
| C3H2BrN3O2 | |
| 1 g | |
| UWRJWMLKEHRGOH-UHFFFAOYSA-N | |
| 2-bromo-5-nitro-1H-imidazole | |
| 47754 | |
| ≥98.0% (GC) | |
| 2-Bromo-4-nitroimidazole |
Safety and Handling
TSCA : No