missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitroimidazole 98.0+%, TCI America™

Catalog No. B35035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35035G 5 g
B35031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35035G Supplier TCI America Supplier No. B35035G

Chemical Identifiers

CAS 65902-59-2
Molecular Formula C3H2BrN3O2
Molecular Weight (g/mol) 191.972
MDL Number MFCD09038915
InChI Key UWRJWMLKEHRGOH-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr
PubChem CID 47754
IUPAC Name 2-bromo-5-nitro-1H-imidazole
SMILES C1=C(NC(=N1)Br)[N+](=O)[O-]

Specifications

Melting Point 235°C
Color Yellow
Quantity 5 g
Formula Weight 191.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitroimidazole