missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. B36075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36075G 5 g
B360725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36075G Supplier TCI America Supplier No. B36075G
Only null left

Chemical Identifiers

CAS 16426-64-5
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
InChI Key CEXGTXNIIFSPSF-UHFFFAOYSA-N
PubChem CID 313534
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 169°C
Color Yellow
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitrobenzoic Acid