missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitroanisole 98.0+%, TCI America™

Catalog No. B28305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28305G 5 g
B283025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28305G Supplier TCI America Supplier No. B28305G

Chemical Identifiers

CAS 5197-28-4
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00041242
InChI Key JMUDXQVNBZCQRF-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroanisole, 3-bromo-4-methoxynitrobenzene, anisole, 2-bromo-4-nitro, benzene, 2-bromo-1-methoxy-4-nitro, 2-bromo-1-methoxy-4-nitro-benzene, pubchem4133, 2-bromo4-nitroanisole, 4-nitro-2-bromanisol, acmc-1awkm, 2-bromo-4-nitro anisole
PubChem CID 78870
IUPAC Name 2-bromo-1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1Br)[N+]([O-])=O

Specifications

Melting Point 104°C
Quantity 5 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitroanisole