missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitroaniline 98.0+%, TCI America™

$78.29 - $242.33

Chemical Identifiers

CAS 13296-94-1
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD00025152
InChI Key CGPPWNTVTNCHDO-UHFFFAOYSA-N
PubChem CID 25840
IUPAC Name 2-bromo-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48731G
View Documents
TCI America
B48731G
1 g
Each for $78.29
Only null left
 
B48735G
View Documents
TCI America
B48735G
5 g
Each for $242.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13296-94-1
217.022
CGPPWNTVTNCHDO-UHFFFAOYSA-N
2-bromo-4-nitroaniline
C6H5BrN2O2
MFCD00025152
25840
C1=CC(=C(C=C1[N+](=O)[O-])Br)N

Specifications

13296-94-1
Yellow
MFCD00025152
CGPPWNTVTNCHDO-UHFFFAOYSA-N
2-bromo-4-nitroaniline
25840
≥98.0% (GC)
2-Bromo-4-nitroaniline
106°C
C6H5BrN2O2
1 g
C1=CC(=C(C=C1[N+](=O)[O-])Br)N
217.022
217.02
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0409

RTECSNumber : BW9350000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'