missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitroaniline 98.0+%, TCI America™

Catalog No. B48731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48731G 1 g
B48735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48731G Supplier TCI America Supplier No. B48731G
Only null left

Chemical Identifiers

CAS 13296-94-1
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD00025152
InChI Key CGPPWNTVTNCHDO-UHFFFAOYSA-N
PubChem CID 25840
IUPAC Name 2-bromo-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)N

Specifications

Melting Point 106°C
Color Yellow
Quantity 1 g
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitroaniline
EINECSNumber (3)-0409
RTECSNumber BW9350000
TSCA Yes