missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(methylsulfonyl)acetophenone 98.0+%, TCI America™

Catalog No. p-7119691
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25735G 5 g
B257325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25735G Supplier TCI America Supplier No. B25735G

Chemical Identifiers

CAS 50413-24-6
Molecular Formula C9H9BrO3S
Molecular Weight (g/mol) 277.13
MDL Number MFCD00673134
InChI Key JOCMYOUZIDSYFO-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methylsulfonyl phenyl-1-ethanone, 2-bromo-1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonyl alpha-bromoacetophenone, 2-bromo-1-4-methylsulfonylphenyl ethanone, 2-bromo-1-4-methanesulfonylphenyl ethan-1-one, 2-bromo-4'-methylsulfonyl acetophenone, 2-bromo-1-4-methanesulfonylphenyl ethanone, 2-bromo-1-4-methanesulfonyl-phenyl-ethanone, ethanone, 2-bromo-1-4-methylsulfonyl phenyl
PubChem CID 735823
IUPAC Name 2-bromo-1-(4-methanesulfonylphenyl)ethan-1-one
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 277.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-(methylsulfonyl)acetophenone
EINECSNumber (3)-4356
TSCA No