accessibility menu, dialog, popup

UserName

CH3SOOOBr

CAS RN 50413-24-6

IUPAC Name: 2-bromo-1-(4-methanesulfonylphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-methanesulfonylphenyl)ethanone
Molecular Weight (g/mol): 277.13
Molecular Formula: C9H9BrO3S
InChi Key: JOCMYOUZIDSYFO-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C(=O)CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results