missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylphenol, 97%

Catalog No. p-7025955
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2170106 5 g
AAB2170114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2170106 Supplier Thermo Scientific Chemicals Supplier No. B2170106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6627-55-0 | C7H7BrO | 187.036 g/mol

It is applied as a compound responsible for iodine off-flavor in wines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a compound responsible for iodine off-flavor in wines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6627-55-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00002151
InChI Key MTIDYGLTAOZOGU-UHFFFAOYSA-N
Synonym 2-bromo-p-cresol, phenol, 2-bromo-4-methyl, 3-bromo-4-hydroxytoluene, 2-bromo-4-methyl-phenol, p-cresol, 2-bromo, pubchem4096, acmc-209nul, 4-06-00-02143 beilstein handbook reference, ksc494c6f
PubChem CID 23109
IUPAC Name 2-bromo-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)Br

Specifications

Melting Point 16°C
Density 1.547
Boiling Point 213°C to 214°C
Flash Point >110°C (230°F)
Odor Phenol-like
Refractive Index 1.578
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylphenol
Show More Show Less
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 229-595-7
RTECSNumber GO6868200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only