missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. B38795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38795G 5 g
B38791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38795G Supplier TCI America Supplier No. B38795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-27-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00045797
InChI Key ZZYYOHPHSYCHQG-UHFFFAOYSA-N
Synonym 4-methyl-2-bromobenzoic acid, 2-bromo-p-toluic acid, 2-bromo-4-methyl-benzoic acid, benzoic acid, 2-bromo-4-methyl, 2-bromo-4-methyl benzoic acid, zlchem 427, pubchem4719, 3-bromo-4-carboxytoluene, acmc-1bbb4, 2-bromo-4-methylbenzoicacid
PubChem CID 226292
IUPAC Name 2-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(Br)=C1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-methylbenzoic Acid