missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylbenzoic acid, 97%

Catalog No. p-7040600
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6041303 1 g
AAH6041306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6041303 Supplier Thermo Scientific Chemicals Supplier No. H6041303

CAS: 7697-27-0 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7697-27-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00045797
InChI Key ZZYYOHPHSYCHQG-UHFFFAOYSA-N
Synonym 4-methyl-2-bromobenzoic acid, 2-bromo-p-toluic acid, 2-bromo-4-methyl-benzoic acid, benzoic acid, 2-bromo-4-methyl, 2-bromo-4-methyl benzoic acid, zlchem 427, pubchem4719, 3-bromo-4-carboxytoluene, acmc-1bbb4, 2-bromo-4-methylbenzoicacid
PubChem CID 226292
IUPAC Name 2-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(Br)=C1

Specifications

Melting Point 143°C to 147°C
Quantity 1 g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylbenzoic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only