missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylaniline 98.0+%, TCI America™

Catalog No. p-7118509
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B05405G 5 g
B054025G 25 g
2 options

Catalog No. B05405G

Supplier: TCI America B05405G

Only null left

Chemical Identifiers

CAS 583-68-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00007635
InChI Key UVRRJILIXQAAFK-UHFFFAOYSA-N
Synonym 2-bromo-p-toluidine, benzenamine, 2-bromo-4-methyl, 4-amino-3-bromotoluene, 2-bromo-4-methylbenzenamine, p-toluidine, 2-bromo, 2-bromo-4-methyl-phenylamine, 3-bromo-4-aminotoluene, 4-methyl-2-bromoaniline, 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline
PubChem CID 11422
IUPAC Name 2-bromo-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Br

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
UN Number 2810
Formula Weight 186.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-methylaniline
RTECSNumber XU4300000
TSCA Yes