Learn More
2-Bromo-4-methylaniline, 99%
Description
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 14°C to 16°C |
| Density | 1.494 |
| Boiling Point | 240°C to 242°C |
| Flash Point | >110°C (230°F) |
| Odor | Characteristic |
| Refractive Index | 1.603 |
| Quantity | 10 g |
| UN Number | UN2810 |
| Beilstein | 1931711 |
| Solubility Information | Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H315-H319-H335-H373 |
| Hazard Statement | GHS H Statement H301-H311-H315-H319 Toxic if swallowed. Toxic in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 209-515-7 |
| RTECSNumber | XU4300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only